Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007507.2 | NP_031533.2 | 3, 4 | 214 | ||
GenBank mRNA | BC028438.1 | - | 3, 4 | 214 | 1332765 | |
BC089550.1 | - | 3, 4 | 206 | 30380642 | ||
S52977.1 | - | 3, 4 | 197 |