Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx3-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007524.3 | NP_031550.2 | 2 | 1174 | ||
GenBank mRNA | BC021014.1 | - | 2 | 1457 | ||
BC145872.1 | - | 2 | 1169 | 40131175 | ||
BC145874.1 | - | 2 | 1169 | 40131177 | ||
U87957.1 | - | 2 | 1174 |