Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007525.3 | NP_031551.1 | 4 | 1083 | ||
GenBank mRNA | AF057157.1 | - | 4 | 1029 | ||
AK145528.1 | - | 4 | 1069 | |||
BC055362.1 | - | 4 | 1016 | |||
BC128370.1 | - | 4 | 947 | 40111665 | ||
BC128371.1 | - | 4 | 947 | 40111666 |