Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013800.2 | NP_038828.2 | 1 | 299 | ||
GenBank mRNA | BC012684.1 | - | 1 | 287 | 3980774 | |
L77900.1 | - | 1 | 97 |