Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009775.4 | NP_033905.3 | 2 | 216 | ||
GenBank mRNA | AA472650.1 | - | 2 | 215 | ||
AK149743.1 | - | 2 | 191 | |||
AK150516.1 | - | 2 | 188 | |||
AK151421.1 | - | 2 | 190 | |||
AK151569.1 | - | 2 | 197 | |||
AK151824.1 | - | 2 | 197 | |||
AK152082.1 | - | 2 | 188 | |||
AK152827.1 | - | 2 | 188 | |||
AK152860.1 | - | 2 | 190 | |||
AK153159.1 | - | 2 | 190 | |||
AK153273.1 | - | 2 | 200 | |||
AK156489.1 | - | 2 | 191 | |||
AK165450.1 | - | 2 | 190 | |||
AK165822.1 | - | 2 | 197 | |||
AK166125.1 | - | 2 | 197 | |||
AK172384.1 | - | 2 | 191 | |||
D21207.1 | - | 2 | 180 | |||
L17306.1 | - | 1 | 145 |