Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009787.2 | NP_033917.2 | 8 | 1397 | ||
GenBank mRNA | AK028292.1 | - | 8 | 1247 | ||
AK146288.1 | - | 8 | 1230 | |||
AK148801.1 | - | 8 | 1319 | |||
AK150566.1 | - | 8 | 1278 | |||
AK161534.1 | - | 8 | 1235 | |||
AK168447.1 | - | 8 | 1201 | |||
AK169387.1 | - | 8 | 1242 | |||
BC066857.1 | - | 7 | 996 | |||
BC141078.1 | - | 8 | 1293 | 9055690 | ||
J05186.1 | - | 8 | 1280 | |||
Y00884.1 | - | 7 | 1103 |