Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243064.1 | NP_001229993.1 | 1 | 322 | ||
NM_007616.4 | NP_031642.1 | 2 | 226 | |||
GenBank mRNA | AB029929.1 | - | 2 | 191 | ||
AB029930.1 | - | 1 | 239 | |||
AK003489.1 | - | 2 | 174 | |||
AK028738.1 | - | 2 | 160 | |||
AK035592.1 | - | 2 | 164 | |||
AK090074.1 | - | 1 | 242 | |||
AY439333.1 | - | 2 | 145 | |||
BC038280.1 | - | 2 | 134 | 4484857 | ||
BC052859.1 | - | 2 | 137 | 30062454 | ||
BY235137.1 | - | 2 | 201 | |||
CK622116.1 | - | 1 | 322 | |||
U07645.1 | - | 2 | 111 |