Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007621.2 | NP_031647.1 | 7 | 636 | ||
GenBank mRNA | AK081816.1 | - | 7 | 607 | ||
BC010758.1 | - | 7 | 596 | 4219349 | ||
BF730701.1 | - | 2 | 382 | |||
D26123.1 | - | 7 | 623 | |||
X07411.1 | - | 7 | 593 |