Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243769.1 | NP_001230698.1 | 5 | 564 | ||
NM_009861.3 | NP_033991.1 | 5 | 564 | |||
GenBank mRNA | AK003098.1 | - | 5 | 485 | ||
AK051543.1 | - | 5 | 485 | |||
AK144216.1 | - | 5 | 487 | |||
AK145659.1 | - | 4 | 328 | |||
AK150563.1 | - | 5 | 497 | |||
AK151087.1 | - | 5 | 509 | |||
AK151726.1 | - | 5 | 486 | |||
AK153564.1 | - | 5 | 508 | |||
AK154870.1 | - | 5 | 483 | |||
AK159470.1 | - | 5 | 526 | |||
AK166281.1 | - | 5 | 483 | |||
AK167195.1 | - | 5 | 472 | |||
AK167400.1 | - | 5 | 477 | |||
AK167609.1 | - | 5 | 482 | |||
AK168013.1 | - | 4 | 477 | |||
AK168076.1 | - | 5 | 477 | |||
AK168089.1 | - | 4 | 477 | |||
AK168276.1 | - | 5 | 477 | |||
AK168503.1 | - | 5 | 498 | |||
AK168758.1 | - | 5 | 517 | |||
AK168820.1 | - | 5 | 490 | |||
AK169122.1 | - | 5 | 498 | |||
AK169232.1 | - | 5 | 477 | |||
AK169805.1 | - | 5 | 502 | |||
AK213611.1 | - | 4 | 338 | |||
BC064792.1 | - | 5 | 470 | 30105331 | ||
CD353254.1 | - | 2 | 148 | |||
L11318.1 | - | 4 | 400 | |||
L78075.1 | - | 4 | 404 | |||
U37720.1 | - | 5 | 472 |