Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009889.2 | NP_034019.1 | 3 | 319 | ||
GenBank mRNA | AF307151.1 | - | 2 | 233 | ||
BC087926.1 | - | 3 | 317 | 30251641 | ||
J00643.1 | - | 3 | 287 |