Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007834.3 | NP_031860.1 | 5 | 699 | ||
GenBank mRNA | AB001990.1 | - | 5 | 514 | ||
AK077485.1 | - | 5 | 513 | |||
AK077628.1 | - | 5 | 505 | |||
AK178078.1 | - | 3 | 234 | |||
AK179221.1 | - | 3 | 234 | |||
AK184129.1 | - | 3 | 235 | |||
AK201554.1 | - | 2 | 89 | |||
AK216820.1 | - | 4 | 355 | |||
BC003740.1 | - | 5 | 487 | 3592608 |