Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007840.3 | NP_031866.2 | 7 | 856 | ||
GenBank mRNA | AK044792.1 | - | 7 | 856 | ||
AK076018.1 | - | 7 | 857 | |||
AK088887.1 | - | 7 | 857 | |||
AK129022.1 | - | 7 | 856 | |||
AK132779.1 | - | 7 | 903 | |||
AK135173.1 | - | 7 | 857 | |||
AK166045.1 | - | 7 | 857 | |||
AK166698.1 | - | 8 | 989 | |||
BC046554.1 | - | 7 | 834 | |||
BC062916.1 | - | 7 | 841 | 6827275 | ||
BC086320.1 | - | 7 | 779 | |||
BC108369.1 | - | 7 | 701 | 6312791 | ||
BC129873.1 | - | 7 | 734 | 40110366 | ||
BC129874.1 | - | 7 | 737 | 40110367 | ||
X65627.1 | - | 7 | 838 |