Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twist2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007855.2 | NP_031881.1 | 1 | 522 | ||
GenBank mRNA | AK008388.1 | - | 1 | 260 | ||
AK008411.1 | - | 1 | 259 | |||
AK011180.1 | - | 1 | 538 | |||
AK011861.1 | - | 1 | 539 | |||
BC090636.1 | - | 1 | 527 | 30942245 | ||
U36384.1 | - | 1 | 522 |