Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039520.2 | NP_001034609.1 | 8 | 1274 | ||
NM_001253893.1 | NP_001240822.1 | 8 | 1274 | |||
NM_001253894.1 | NP_001240823.1 | 8 | 1274 | |||
NM_007871.2 | NP_031897.2 | 8 | 1274 | |||
GenBank mRNA | AK005012.2 | - | 8 | 1250 | ||
AK015410.1 | - | 7 | 939 | |||
AK150046.1 | - | 8 | 1237 | |||
AK154375.1 | - | 7 | 938 | |||
AK157844.1 | - | 8 | 1244 | |||
AK170574.1 | - | 8 | 1250 | |||
AK171049.1 | - | 8 | 1228 | |||
AK171566.1 | - | 8 | 1238 | |||
AK172231.1 | - | 8 | 1255 | |||
AK208713.1 | - | 2 | 111 | |||
L31398.1 | - | 8 | 1214 |