Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010069.1 | NP_034199.1 | 11 | 1771 | ||
GenBank mRNA | BC055768.1 | - | 11 | 1771 | 6413208 | |
D50000.1 | - | 10 | 1194 |
Gene Symbol: Ino80e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135531.1 | - | 7 | 1621 | ||
BC025527.1 | - | 6 | 1971 |