Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aimp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007926.2 | NP_031952.2 | 2 | 168 | ||
GenBank mRNA | AK087932.1 | - | 2 | 123 | ||
AK133859.1 | - | 2 | 121 | |||
AK160506.1 | - | 2 | 119 | |||
BC002054.1 | - | 2 | 123 | 3496254 | ||
BY256548.1 | - | 2 | 173 | |||
U10118.1 | - | 2 | 119 |