Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007929.2 | NP_031955.2 | 3, 4 | 417 | ||
GenBank mRNA | AF083876.1 | - | 3, 4 | 284 | ||
AF346627.1 | - | 3, 4 | 359 | |||
AK076194.1 | - | 3, 4 | 396 | |||
AK077260.1 | - | 3, 4 | 419 | |||
BC031646.1 | - | 2, 3 | 210 | |||
BC041670.1 | - | 3, 4 | 320 | 3663158 |