Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080387.2 | NP_001073856.1 | 5 | 631 | ||
NM_001284195.1 | NP_001271124.1 | 5 | 628 | |||
NM_001284196.1 | NP_001271125.1 | 5 | 628 | |||
NM_010178.3 | NP_034308.1 | 5 | 631 | |||
GenBank mRNA | AB015894.1 | - | 5 | 542 | ||
AB015895.1 | - | 5 | 539 | |||
AF042383.1 | - | 5 | 543 | |||
AF060490.1 | - | 5 | 566 | |||
AK014345.1 | - | 5 | 570 | |||
AK134076.1 | - | 4 | 921 | |||
AK134289.1 | - | 5 | 601 | |||
AK135316.1 | - | 5 | 576 | |||
AK150734.1 | - | 5 | 565 | |||
AK151567.1 | - | 5 | 570 | |||
AK168524.1 | - | 5 | 589 | |||
AK168558.1 | - | 5 | 589 | |||
AK168571.1 | - | 5 | 554 | |||
AK169071.1 | - | 5 | 555 | |||
BC006670.1 | - | 5 | 583 | |||
BC037591.1 | - | 5 | 549 | 5004611 | ||
BC043060.1 | - | 5 | 955 | 6419883 | ||
BC083082.1 | - | 5 | 543 | 6511807 | ||
CF914989.1 | - | 5 | 544 | |||
CN528501.1 | - | 4 | 330 |