Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111066.1 | NP_001104536.1 | 8 | 1131 | ||
NM_001199631.1 | NP_001186560.1 | 8 | 1227 | |||
NM_010223.2 | NP_034353.2 | 8 | 1128 | |||
GenBank mRNA | AB154247.1 | - | 7 | 867 | ||
AF030635.1 | - | 7 | 898 | |||
AK088739.1 | - | 8 | 1066 | |||
AK146122.1 | - | 8 | 1126 | |||
AK152198.1 | - | 8 | 1127 | |||
AK153000.1 | - | 8 | 1124 | |||
AK165281.1 | - | 8 | 1124 | |||
AK167663.1 | - | 8 | 1114 | |||
AK189147.1 | - | 2 | 109 | |||
AY225340.1 | - | 7 | 1011 | |||
AY278608.1 | - | 8 | 1087 | |||
BC003739.1 | - | 7 | 1047 | 3601511 | ||
BC027808.1 | - | 8 | 1081 | 5324380 |