Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gas6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019521.2 | NP_062394.2 | 3, 4 | 465 | ||
GenBank mRNA | AK086187.1 | - | 3, 4 | 459 | ||
BC005444.1 | - | 3, 4 | 420 | 3592398 | ||
X59846.1 | - | 3, 4 | 436 |