Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271002.1 | NP_001257931.1 | 1, 2 | 73 | ||
NM_020014.2 | NP_064398.1 | 1, 2 | 73 | |||
GenBank mRNA | AJ276514.1 | - | 1, 2 | 73 | ||
AJ276515.1 | - | 1, 2 | 73 | |||
AJ276516.1 | - | 1, 2 | 73 | |||
BC111808.1 | - | 1, 2 | 72 | |||
BC141873.1 | - | 1, 2 | 68 | |||
BC141874.1 | - | 1, 2 | 73 | 40040202 |