Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008167.2 | NP_032193.1 | 3 | 492 | ||
GenBank mRNA | AK036192.1 | - | 3 | 523 | ||
AK141070.1 | - | 1344 | ||||
BC139823.1 | - | 3 | 496 | 40141838 | ||
D13266.1 | - | 3 | 492 |