Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029850.4 | NP_001025021.1 | 12 | 2462 | ||
NM_001083320.2 | NP_001076789.1 | 11 | 1804 | |||
NM_001083321.2 | NP_001076790.1 | 11 | 1804 | |||
NM_001286784.1 | NP_001273713.1 | 11 | 2426 | |||
NM_001286785.1 | NP_001273714.1 | 12 | 2462 | |||
NM_001286786.1 | NP_001273715.1 | 11 | 2426 | |||
NM_001286788.1 | NP_001273717.1 | 11 | 1804 | |||
NM_010367.3 | NP_034497.1 | 11 | 2426 | |||
GenBank mRNA | AB194411.1 | - | 11 | 1831 | ||
AB194412.1 | - | 11 | 1831 | |||
AF027503.1 | - | 11 | 2755 | |||
AK147358.1 | - | 12 | 2463 | |||
AK147666.1 | - | 11 | 1519 | |||
AK163582.1 | - | 8 | 1129 | |||
AK163800.1 | - | 11 | 1669 | |||
AK220499.1 | - | 11 | 1509 | |||
AY497557.1 | - | 12 | 1867 | |||
BC060273.1 | - | 11 | 1804 | |||
BC095943.1 | - | 12 | 2440 | |||
BC150820.1 | - | 12 | 2136 | 9087731 |