Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032572.1 | - | 12 | 3159 | ||
AK143616.1 | - | 16 | 2090 | |||
AK144669.1 | - | 16 | 2100 | |||
AK146813.1 | - | 16 | 2062 | |||
AK146918.1 | - | 16 | 2067 | |||
AK214335.1 | - | 3 | 252 | |||
AK217149.1 | - | 1 | 15 | |||
BC003741.1 | - | 16 | 2059 | 3488803 | ||
D63850.1 | - | 16 | 2076 | |||
FN687734.2 | - | 16 | 1816 |