Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032572.1 | - | 4, 5 | 687 | ||
AK143616.1 | - | 4, 5 | 608 | |||
AK144669.1 | - | 4, 5 | 612 | |||
AK146813.1 | - | 4, 5 | 547 | |||
AK146918.1 | - | 4, 5 | 585 | |||
AK186839.1 | - | 1, 2 | 22 | |||
AK208959.1 | - | 1, 2 | 22 | |||
AK214884.1 | - | 1, 2 | 22 | |||
BC003741.1 | - | 4, 5 | 544 | 3488803 | ||
D63850.1 | - | 4, 5 | 588 | |||
FN687734.2 | - | 4, 5 | 331 |