Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010414.3 | NP_034544.1 | 3, 4 | 579 | ||
GenBank mRNA | AK042204.1 | - | 3, 4 | 580 | ||
L28827.1 | - | 3, 4 | 498 | |||
U24233.1 | - | 3, 4 | 539 |