Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010415.2 | NP_034545.1 | 1 | 313 | ||
GenBank mRNA | AK052084.1 | - | 1 | 284 | ||
AK145548.1 | - | 1 | 282 | |||
AK160575.1 | - | 1 | 283 | |||
AK164309.1 | - | 1 | 314 | |||
BC089607.1 | - | 1 | 282 | 6751441 | ||
L07264.1 | - | 1 | 246 |