Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfb2m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008249.4 | NP_032275.2 | 2 | 497 | ||
GenBank mRNA | AK090106.1 | - | 2 | 468 | ||
AK146563.1 | - | 2 | 498 | |||
AK166717.1 | - | 2 | 484 | |||
AK167930.1 | - | 2 | 449 | |||
AK168970.1 | - | 2 | 476 | |||
AK169138.1 | - | 2 | 476 | |||
AK210035.1 | - | 1 | 62 | |||
BC138836.1 | - | 2 | 442 | 8861858 | ||
BC138837.1 | - | 2 | 442 | 8861859 | ||
M74555.1 | - | 2 | 448 |