Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010441.2 | NP_034571.1 | 3, 4 | 1116 | ||
GenBank mRNA | AK028346.1 | - | 3, 4 | 446 | ||
AK142059.1 | - | 3, 4 | 478 | |||
BC052158.1 | - | 3, 4 | 650 | 6506864 | ||
BC069985.1 | - | 3, 4 | 344 | 6332337 | ||
BC085085.1 | - | 3, 4 | 417 | 6334495 | ||
X58380.1 | - | 3, 4 | 330 |
Gene Symbol: Hmga2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033703.1 | - | 1 | 152 |