Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010450.3 | NP_034580.1 | 1 | 178 | ||
GenBank mRNA | AK133481.1 | - | 1 | 178 | ||
BC052232.1 | - | 1 | 156 | |||
BC119302.1 | - | 1 | 113 | 8734051 | ||
U20370.1 | - | 1 | 134 |
Gene Symbol: Hoxa11os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | U20366.1 | - | LIT1565 Hoxa11as |
1 | 387 | |
U20367.1 | - | LIT1564 Hoxa11as |
1 | 548 |