Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Id2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010496.3 | NP_034626.1 | 2 | 453 | ||
GenBank mRNA | AK003222.1 | - | 2 | 450 | ||
AK013239.1 | - | 1 | 736 | |||
AK013378.1 | - | 2 | 450 | |||
AK135547.1 | - | 2 | 455 | |||
AK146346.1 | - | 2 | 453 | |||
AK146606.1 | - | 2 | 456 | |||
BC006921.1 | - | 2 | 430 | 2655173 | ||
BC053699.1 | - | 2 | 440 | 6515664 | ||
M69293.1 | - | 2 | 447 |