Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008342.3 | NP_032368.2 | 1, 2 | 479 | ||
GenBank mRNA | AK011784.1 | - | 2, 3 | 350 | ||
AK011790.1 | - | 1, 2 | 408 | |||
AK012703.1 | - | 1, 2 | 469 | |||
BC012724.1 | - | 1, 2 | 467 | 2654907 | ||
BC054473.1 | - | 1, 2 | 452 | 6488878 | ||
X81580.1 | - | 1, 2 | 450 |