Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008449.2 | NP_032475.2 | 10 | 1195 | ||
GenBank mRNA | AB093244.1 | - | 10 | 1088 | ||
AF067180.1 | - | 10 | 847 | |||
AK158884.1 | - | 10 | 1181 | |||
BC067051.1 | - | 10 | 1195 | 30536079 |