Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008449.2 | NP_032475.2 | 24 | 2983 | ||
GenBank mRNA | AB093244.1 | - | 24 | 2876 | ||
AF067180.1 | - | 24 | 2635 | |||
AK158884.1 | - | 24 | 2969 | |||
BC067051.1 | - | 24 | 2983 | 30536079 |