Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010694.1 | NP_034824.1 | 3 | 252 | ||
GenBank mRNA | BC116816.1 | - | 3 | 273 | 40126135 | |
BC116818.1 | - | 3 | 273 | 40126137 | ||
BC144882.1 | - | 3 | 273 | |||
D38580.1 | - | 3 | 252 |