Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136069.2 | NP_001129541.2 | 8 | 1180 | ||
NM_010699.2 | NP_034829.1 | 8 | 987 | |||
GenBank mRNA | AK089929.1 | - | 8 | 951 | ||
AK146121.1 | - | 8 | 948 | |||
AK146980.1 | - | 4 | 362 | |||
AK150156.1 | - | 8 | 951 | |||
AK167789.1 | - | 8 | 949 | |||
AK167944.1 | - | 8 | 949 | |||
AK168346.1 | - | 8 | 948 | |||
AK168920.1 | - | 8 | 953 | |||
AK170386.1 | - | 8 | 952 | |||
AK170708.1 | - | 8 | 949 | |||
AK171373.1 | - | 7 | 832 | |||
AK200149.1 | - | 4 | 296 | |||
AK206536.1 | - | 2 | 45 | |||
AK207234.1 | - | 2 | 45 | |||
AK207750.1 | - | 2 | 45 | |||
AK207846.1 | - | 2 | 45 | |||
AK212267.1 | - | 2 | 45 | |||
AK217640.1 | - | 2 | 45 | |||
AK219220.1 | - | 2 | 45 | |||
BC005509.1 | - | 7 | 799 | |||
BC066858.1 | - | 7 | 605 | |||
BC094019.1 | - | 8 | 904 | 4195220 | ||
BC094428.1 | - | 8 | 941 | 30700835 | ||
M17516.1 | - | 3 | 246 | |||
U13687.1 | - | 8 | 1047 |