Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013587.2 | NP_038615.2 | 7, 8 | 1029 | ||
GenBank mRNA | AJ639614.1 | - | 7, 8 | 1036 | ||
AK089250.1 | - | 7, 8 | 1032 | |||
AK145710.1 | - | 7, 8 | 1072 | |||
AK150451.1 | - | 7, 8 | 1042 | |||
AK152062.1 | - | 1, 2 | 91 | |||
AK152485.1 | - | 7, 8 | 1042 | |||
AK153323.1 | - | 7, 8 | 1042 | |||
AK166616.1 | - | 7, 8 | 1032 | |||
AK166746.1 | - | 7, 8 | 1043 | |||
AK166819.1 | - | 7, 8 | 1047 | |||
AK168421.1 | - | 7, 8 | 1032 | |||
AK169132.1 | - | 7, 8 | 1037 | |||
AK171471.1 | - | 7, 8 | 1032 | |||
BC032170.1 | - | 7, 8 | 1032 | |||
BC046641.1 | - | 7, 8 | 1021 | |||
BC057979.1 | - | 7, 8 | 1000 | |||
BC059887.1 | - | 7, 8 | 846 | |||
BC094324.1 | - | 7, 8 | 1029 | 6402163 | ||
D00622.1 | - | 7, 8 | 1022 | |||
S67967.1 | - | 7, 8 | 1055 |