Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013587.2 | NP_038615.2 | 4 | 540 | ||
GenBank mRNA | AJ639614.1 | - | 4 | 547 | ||
AK089250.1 | - | 4 | 543 | |||
AK145710.1 | - | 4 | 583 | |||
AK150451.1 | - | 4 | 554 | |||
AK152485.1 | - | 4 | 554 | |||
AK153323.1 | - | 4 | 554 | |||
AK166616.1 | - | 4 | 543 | |||
AK166746.1 | - | 4 | 554 | |||
AK166819.1 | - | 4 | 558 | |||
AK168421.1 | - | 4 | 543 | |||
AK169132.1 | - | 4 | 548 | |||
AK171471.1 | - | 4 | 543 | |||
AK206813.1 | - | 2 | 76 | |||
BC032170.1 | - | 4 | 543 | |||
BC046641.1 | - | 4 | 532 | |||
BC057979.1 | - | 4 | 511 | |||
BC059887.1 | - | 4 | 357 | |||
BC094324.1 | - | 4 | 540 | 6402163 | ||
D00622.1 | - | 4 | 533 | |||
S67967.1 | - | 4 | 566 |