Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008520.2 | NP_032546.2 | 12, 13 | 2708 | ||
GenBank mRNA | AK038587.1 | - | 9, 10 | 1645 | ||
AK080869.1 | - | 12, 13 | 2708 | |||
AK131126.1 | - | 8, 9 | 3247 | |||
BC127051.1 | - | 11, 12 | 1307 | |||
L40459.1 | - | 11, 12 | 1824 |