Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Creld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133930.1 | NP_598691.1 | 6 | 1018 | ||
GenBank mRNA | AB009020.1 | - | 6 | 1023 | ||
AK077726.1 | - | 6 | 1163 | |||
AK155777.1 | - | 6 | 1028 | |||
AK181275.1 | - | 1 | 27 | |||
BC010804.1 | - | 6 | 1018 | 4239786 | ||
BC023893.1 | - | 6 | 1029 | 5310111 | ||
BC025932.1 | - | 6 | 924 | 5102571 | ||
BC029065.1 | - | 6 | 1054 | 4935296 |