Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008570.1 | NP_032596.1 | 3 | 271 | ||
GenBank mRNA | BC100700.1 | - | 3 | 262 | 40043851 | |
BC100701.1 | - | 3 | 262 | 40043850 | ||
BC100702.1 | - | 3 | 262 | 40043849 | ||
BC100703.1 | - | 3 | 262 | 40043848 | ||
S44609.1 | - | 3 | 264 |