Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109973.2 | NP_001103443.1 | 3 | 520 | ||
NM_001276390.1 | NP_001263319.1 | 2 | 402 | |||
NM_001276391.1 | NP_001263320.1 | 4 | 292 | |||
NM_010783.3 | NP_034913.2 | 3 | 188 | |||
GenBank mRNA | AA048074.1 | - | 3 | 257 | ||
BC006018.1 | - | 2 | 155 | 3488088 | ||
BC010259.1 | - | 3 | 360 | 3481922 | ||
BC083058.1 | - | 3 | 293 | 5354539 | ||
BC085233.1 | - | 3 | 520 | 5318215 | ||
BQ832230.1 | - | 2 | 369 | |||
CX564914.1 | - | 3 | 259 | |||
U57820.1 | - | 2 | 160 | |||
U57821.1 | - | 3 | 158 | |||
U57822.1 | - | 2 | 340 |