Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 14 | 1973 | ||
NM_018752.3 | NP_061222.3 | 14 | 1866 | |||
GenBank mRNA | AF047714.1 | - | 14 | 1735 | ||
AK044507.1 | - | 14 | 1897 | |||
AK148193.1 | - | 12 | 1815 | |||
AK148268.1 | - | 14 | 1720 | |||
AK148275.1 | - | 14 | 1776 | |||
AK149290.1 | - | 14 | 1895 | |||
AK196486.1 | - | 4 | 294 | |||
AK217903.1 | - | 4 | 295 | |||
AY180104.2 | - | 14 | 1973 |