Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010858.4 | NP_034988.2 | 6, 7 | 636 | ||
GenBank mRNA | AK013098.1 | - | 4, 5 | 564 | ||
AK084544.1 | - | 1, 2 | 2585 | |||
BC048485.1 | - | 6, 7 | 636 | 6741975 | ||
M19436.1 | - | 2, 3 | 318 |