Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myt1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093775.1 | NP_001087244.1 | 11 | 2447 | ||
NM_001093776.1 | NP_001087245.1 | 11 | 2353 | |||
NM_001093778.1 | NP_001087247.1 | 11 | 2438 | |||
NM_008666.3 | NP_032692.2 | 11 | 2441 | |||
GenBank mRNA | AB093283.1 | - | 8 | 1800 | ||
AB212898.1 | - | 10 | 2045 | |||
AF004295.1 | - | 11 | 2260 | |||
BC079651.1 | - | 11 | 2322 | |||
BC094438.1 | - | 7 | 1678 | 30692780 | ||
BC131677.1 | - | 10 | 2042 | 40111036 | ||
CD802997.1 | - | 2 | 509 | |||
U86338.1 | - | 11 | 2169 |