Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFATC2 SIRNA Assays ›
Gene Symbol:
Nfatc2
Gene Name:
nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 2
Gene Aliases:
AI607462, NF-ATc2, NF-ATp, NFAT1, NFAT1-D, Nfatp
Chromosome Location:
Chr.2: 168476410 - 168601657 on Build GRCm38
UniGene ID:
Mm.116802
Species:
Mouse
siRNA ID
MSS275984
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1320001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Nfatc2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037177.2
NP_001032254.1
6
1973
NM_001037178.2
NP_001032255.1
6
1338
NM_001136073.2
NP_001129545.1
6
1973
NM_001291168.1
NP_001278097.1
6
1858
NM_001291169.1
NP_001278098.1
6
1858
NM_001291170.1
NP_001278099.1
6
1338
NM_001291171.1
NP_001278100.1
5
822
NM_001291172.1
NP_001278101.1
6
1858
NM_001291173.1
NP_001278102.1
5
822
NM_001291174.1
NP_001278103.1
6
1338
NM_001291175.1
NP_001278104.1
6
1453
NM_001291176.1
NP_001278105.1
5
937
NM_001291177.1
NP_001278106.1
6
1453
NM_001291178.1
NP_001278107.1
5
937
NM_001291179.1
NP_001278108.1
6
1453
NM_010899.3
NP_035029.2
6
1973
GenBank mRNA
AF289078.1
-
6
1799
AK044489.1
-
6
1859
AK083119.1
-
6
1314
AK161174.1
-
4
548
EU887581.1
-
6
1739
EU887582.1
-
6
1799
EU887583.1
-
6
1336
EU887584.1
-
6
1436
EU887585.1
-
5
820
EU887586.1
-
5
920
EU887587.1
-
6
1739
EU887588.1
-
6
1799
EU887589.1
-
6
1336
EU887590.1
-
6
1436
EU887591.1
-
5
820
EU887592.1
-
5
920
EU887599.1
-
6
1739
EU887600.1
-
6
1799
EU887601.1
-
6
1336
EU887602.1
-
6
1436
EU887603.1
-
5
820
EU887604.1
-
5
920
U36575.1
-
6
1869
U36576.1
-
6
1869
Back To Top
More Information
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
positive regulation of gene expression
myotube cell development
cell migration
positive regulation of B cell proliferation
calcineurin-NFAT signaling cascade
response to drug
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
B cell receptor signaling pathway
positive regulation of myoblast fusion
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0