Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122952.1 | NP_001116424.1 | 10 | 1594 | ||
NM_001122953.1 | NP_001116425.1 | 9 | 1704 | |||
NM_010905.3 | NP_035035.1 | 10 | 1833 | |||
GenBank mRNA | AF326553.1 | - | 10 | 1532 | ||
AF326554.1 | - | 10 | 1532 | |||
AK052204.1 | - | 10 | 1655 | |||
AK054310.1 | - | 10 | 1697 | |||
AK137731.1 | - | 10 | 1696 | |||
AK139357.1 | - | 10 | 1595 | |||
BC075702.1 | - | 11 | 1763 | 30087137 | ||
BC122880.1 | - | 9 | 1424 | |||
D90173.1 | - | 10 | 1671 | |||
D90174.1 | - | 10 | 1704 | |||
D90175.1 | - | 1548 | ||||
U57633.1 | - | 10 | 1488 | |||
Y07690.1 | - | 10 | 1564 | |||
Y07691.1 | - | 9 | 1433 |