Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ninj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013610.2 | NP_038638.1 | 3 | 381 | ||
GenBank mRNA | AK077602.1 | - | 3 | 400 | ||
AK155741.1 | - | 3 | 391 | |||
AK159111.1 | - | 3 | 401 | |||
AK160045.1 | - | 3 | 388 | |||
AK171973.1 | - | 3 | 400 | |||
AK183237.1 | - | 1 | 40 | |||
BC049600.1 | - | 3 | 368 | |||
BC118937.1 | - | 3 | 381 | 40091105 | ||
U91513.1 | - | 3 | 381 |