Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008710.3 | NP_032736.2 | 2625 | |||
GenBank mRNA | AK033282.1 | - | 11, 12 | 1813 | ||
AK040460.1 | - | 9, 10 | 1491 | |||
AK050478.1 | - | 10, 11 | 2530 | |||
AK052234.1 | - | 11, 12 | 1857 | |||
AK054332.1 | - | 11, 12 | 1858 | |||
AK083413.1 | - | 11, 12 | 1856 | |||
AK084546.1 | - | 11, 12 | 1855 | |||
AK084604.1 | - | 11, 12 | 1855 | |||
AK085903.1 | - | 9, 10 | 1516 | |||
AK088044.1 | - | 11, 12 | 2607 | |||
AK090115.1 | - | 2628 | ||||
AK129006.1 | - | 11, 12 | 2602 | |||
AK159694.1 | - | 12, 13 | 2611 | |||
AK168736.1 | - | 10, 11 | 2408 | |||
BC008518.1 | - | 11, 12 | 2588 | 3601702 | ||
Z49204.1 | - | 10, 11 | 2532 |