Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139509.1 | NP_001132981.1 | 7 | 1748 | ||
NM_013613.2 | NP_038641.1 | 7 | 1894 | |||
GenBank mRNA | AK087495.1 | - | 8 | 1772 | ||
AK158607.1 | - | 7 | 1895 | |||
BC137715.1 | - | 7 | 2206 | 8860734 | ||
S53744.1 | - | 7 | 1785 |